Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 1M solution in methanol, AcroSeal™

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 428958000
Price (EUR)
346,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
346,00
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration with HCl0.95 to 1.10 M
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only