Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™
Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™
Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™
Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Dilithium tetrachlorocuprate, 0.1M solution in THF, AcroSeal™

CAS: 15489-27-7 | Cl4CuLi2 | 219.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
429051000100 mLAcroSeal™ Glass Bottle
Catalog number 429051000
Price (EUR)
98,65
Online Exclusive
109,00
Save 10,35 (9%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
98,65
Online Exclusive
109,00
Save 10,35 (9%)
Each
Chemical Identifiers
CAS15489-27-7
IUPAC Namecopper(2+) dilithium(1+) tetrachloride
Molecular FormulaCl4CuLi2
InChI KeyHCJWWBBBSCXJMS-UHFFFAOYSA-J
SMILES[Li+].[Li+].[Cl-].[Cl-].[Cl-].[Cl-].[Cu++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear orange to red-brown
Titration Argentometric0.09 to 0.12 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only