3-Bromo-5-hydroxypyridine, 97%
3-Bromo-5-hydroxypyridine, 97%
3-Bromo-5-hydroxypyridine, 97%
Thermo Scientific Chemicals

3-Bromo-5-hydroxypyridine, 97%

CAS: 74115-13-2 | C5H4BrNO | 174.00 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4299000101 gGlass bottle
Catalog number 429900010
Price (EUR)
42,30
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
42,30
Each
Chemical Identifiers
CAS74115-13-2
IUPAC Name5-bromopyridin-3-ol
Molecular FormulaC5H4BrNO
InChI KeyVNYBIBSZZDAEOK-UHFFFAOYSA-N
SMILESOC1=CN=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point167°C to 171°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only