4-Ethynylaniline, 97%
4-Ethynylaniline, 97%
4-Ethynylaniline, 97%
Thermo Scientific Chemicals

4-Ethynylaniline, 97%

CAS: 14235-81-5 | C8H7N | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 432950050
Price (EUR)
206,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
206,00
Each
Chemical Identifiers
CAS14235-81-5
IUPAC Name4-ethynylaniline
Molecular FormulaC8H7N
InChI KeyJXYITCJMBRETQX-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)Yellow to brown
Melting point98°C to 104°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only