N-BOC-Allylamine, 98%
N-BOC-Allylamine, 98%
N-BOC-Allylamine, 98%
Thermo Scientific Chemicals

N-BOC-Allylamine, 98%

CAS: 78888-18-3 | C8H15NO2 | 157.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Catalog number 434770100
Price (EUR)
61,70
Each
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
61,70
Each
Chemical Identifiers
CAS78888-18-3
IUPAC Nametert-butyl N-(prop-2-en-1-yl)carbamate
Molecular FormulaC8H15NO2
InChI KeyAWARHXCROCWEAK-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Low melting solid
Melting point33°C to 40°C
Appearance (Color)Colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only