4-Bromoisoindoline hydrochloride, 97%
4-Bromoisoindoline hydrochloride, 97%
4-Bromoisoindoline hydrochloride, 97%
Thermo Scientific Chemicals

4-Bromoisoindoline hydrochloride, 97%

CAS: 923590-95-8 | C8H8BrN·ClH | 234.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 435870010
Price (EUR)
99,30
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
99,30
Each
Chemical Identifiers
CAS923590-95-8
IUPAC Name4-bromo-2,3-dihydro-1H-isoindole hydrochloride
Molecular FormulaC8H9BrClN
InChI KeyFQHLHVFOJBANKY-UHFFFAOYSA-N
SMILESCl.BrC1=CC=CC2=C1CNC2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Elemental Analysis5.73 to 6.21 % N
Appearance (Color)Off-white to yellow to pink to brown
Infrared spectrumConforms
Elemental Analysis39.33 to 42.61 % C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only