5,5-Dibromobarbituric acid, 95+%
5,5-Dibromobarbituric acid, 95+%
5,5-Dibromobarbituric acid, 95+%
Thermo Scientific Chemicals

5,5-Dibromobarbituric acid, 95+%

CAS: 511-67-1 | C4H2Br2N2O3 | 285.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 439840010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS511-67-1
IUPAC Name5,5-dibromo-1,3-diazinane-2,4,6-trione
Molecular FormulaC4H2Br2N2O3
InChI KeyAMATXUCYHHHHHB-UHFFFAOYSA-N
SMILESBrC1(Br)C(=O)NC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=95.0 %
Infrared spectrumConforms
Loss on drying=<0.5 % (105°C)
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only