2-Fluorophenethyl bromide, 97%
2-Fluorophenethyl bromide, 97%
2-Fluorophenethyl bromide, 97%
Thermo Scientific Chemicals

2-Fluorophenethyl bromide, 97%

CAS: 91319-54-9 | C8H8BrF | 203.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 440120050
Price (EUR)
161,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
161,00
Each
Chemical Identifiers
CAS91319-54-9
IUPAC Name1-(2-bromoethyl)-2-fluorobenzene
Molecular FormulaC8H8BrF
InChI KeyFQGDFWIQBCQXPS-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only