Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Thermo Scientific Chemicals

Propidium iodide, 95%

CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
440301000100 mgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 440301000
Price (EUR)
159,65
Online Exclusive
177,00
Save 17,35 (10%)
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
159,65
Online Exclusive
177,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS25535-16-4
IUPAC Name3,8-diamino-5-{3-[diethyl(methyl)azaniumyl]propyl}-6-phenylphenanthridin-5-ium diiodide
Molecular FormulaC27H34I2N4
InChI KeyXJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES[I-].[I-].CC[N+](C)(CC)CCC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<5 %
Titration Argentometric94 to 106 %
Infrared spectrumConforms
Appearance (Color)Red
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only