Sodium hydroxide, technical, 30% solution in water
Sodium hydroxide, technical, 30% solution in water
Sodium hydroxide, technical, 30% solution in water
Thermo Scientific Chemicals

Sodium hydroxide, technical, 30% solution in water

CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4443500505 LPlastic drum
Catalog number 444350050
Price (EUR)
79,20
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
79,20
Each
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl28.0 to 33.5 %
Appearance (Color)Clear colorless
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only