Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Thermo Scientific Chemicals

Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™

CAS: 373-57-9 | BF3 | 67.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 445278000
Price (EUR)
289,00
Each
Quantity:
800 mL
Request bulk or custom format
Price (EUR)
289,00
Each
Chemical Identifiers
CAS373-57-9
IUPAC Nametrifluoroborane
Molecular FormulaBF3
InChI KeyWTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILESFB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to amber
Appearance (Form)Solution
Assay10 to 15 % w/v (BF3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only