3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%
3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%
Thermo Scientific Chemicals

3-(Tetrahydropyran-2-yloxy)prop-1-en-1-ylboronic acid pinacol ester, 90%

CAS: 642066-70-4 | C14H25BO4 | 268.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 445540050
Price (EUR)
1.203,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
1.203,00
Each
Chemical Identifiers
CAS642066-70-4
IUPAC Name4,4,5,5-tetramethyl-2-[3-(oxan-2-yloxy)prop-1-en-1-yl]-1,3,2-dioxaborolane
Molecular FormulaC14H25BO4
InChI KeyMHSOBXCZCRNELG-UHFFFAOYNA-N
SMILESCC1(C)OB(OC1(C)C)C=CCOC1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Appearance (Color)Colorless
Appearance (Form)Oily liquid
GC>=88.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only