Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™
Thermo Scientific Chemicals

Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 446141000
Price (EUR)
50,60
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (EUR)
50,60
Each
Chemical Identifiers
CAS123-75-1
IUPAC Namepyrrolidine
Molecular FormulaC4H9N
InChI KeyRWRDLPDLKQPQOW-UHFFFAOYSA-N
SMILESC1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.4 %
Water=<0.2 %
Refractive index1.4421 to 1.4441 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only