Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Cyclohexane, ACS reagent
Thermo Scientific Chemicals

Cyclohexane, ACS reagent

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Plastic drum
Catalog number 446280050
Price (EUR)
123,00
Each
Add to cart
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
123,00
Each
Add to cart
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.0 %
Color scale=<10 APHA
Water=<0.02 %
Residue after evaporation=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only