2',7'-Dichlorofluorescein, ACS reagent
2',7'-Dichlorofluorescein, ACS reagent
2',7'-Dichlorofluorescein, ACS reagent
Thermo Scientific Chemicals

2',7'-Dichlorofluorescein, ACS reagent

CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 446930010
Price (EUR)
49,50
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,50
Each
Chemical Identifiers
CAS76-54-0
IUPAC Name2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC20H10Cl2O5
InChI KeyVFNKZQNIXUFLBC-UHFFFAOYSA-N
SMILESOC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light orange to red-brown
Clarity of alcohol solutionPasses test
Appearance (Form)Powder
Suitability(as adsorption indicator) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only