Sodium formate, ACS reagent
Sodium formate, ACS reagent
Sodium formate, ACS reagent
Thermo Scientific Chemicals

Sodium formate, ACS reagent

CAS: 141-53-7 | CHNaO2 | 68.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 446950025
Price (EUR)
202,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
202,00
Each
Chemical Identifiers
CAS141-53-7
IUPAC Namesodium formate
Molecular FormulaCHNaO2
InChI KeyHLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES[Na+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Insoluble matter=<0.005 %
Iron (Fe)=<5 ppm
Titration with KMnO4>=99.0 %
Sulfate (SO4)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only