Potassium phosphate, monobasic, for HPLC
Thermo Scientific Chemicals

Potassium phosphate, monobasic, for HPLC

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 447370500
Price (EUR)
52,65
キャンペーン価格
58,80
Save 6,15 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
52,65
キャンペーン価格
58,80
Save 6,15 (10%)
Each
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.05
Gradient analysispasses test
Titration with NaOH>=99.4 %
UVat 250 nm A: =<0.05
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only