Acetone, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Acetone, specified according to the requirements of Ph.Eur.

CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447450250
Price (EUR)
369,00
Each
Quantity:
25 L
Request bulk or custom format
Price (EUR)
369,00
Each
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Relative density0.790 to 0.793
Related substances=<2 ppm (Benzene)
Insoluble matter(in water) Passes test
Related substances=<0.05 % (Methanol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only