Citric acid, monosodium salt, 99%, for analysis, anhydrous
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Citric acid, monosodium salt, 99%, for analysis, anhydrous
Thermo Scientific Chemicals

Citric acid, monosodium salt, 99%, for analysis, anhydrous

CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 447690010
Price (EUR)
95,30
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
95,30
Each
Chemical Identifiers
CAS18996-35-5
IUPAC Namesodium 3-carboxy-2-(carboxymethyl)-2-hydroxypropanoate
Molecular FormulaC6H7NaO7
InChI KeyHWPKGOGLCKPRLZ-UHFFFAOYSA-M
SMILES[Na+].OC(=O)CC(O)(CC(O)=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
SolubilityClear colorless
Chloride (Cl)=<0.005 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Sulfate (SO4)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only