Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
Benzyl alcohol, ACS reagent
Thermo Scientific Chemicals

Benzyl alcohol, ACS reagent

CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4477200101 LGlass Bottle
Catalog number 447720010
Price (EUR)
132,00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
132,00
Each
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=99.0 %
GC=<0.02 % Acetophenone
GC=<0.01 % Benzaldehyde
Ash=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only