Potassium nitrate, 99+%, for biochemistry
Potassium nitrate, 99+%, for biochemistry
Potassium nitrate, 99+%, for biochemistry
Thermo Scientific Chemicals

Potassium nitrate, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
447825000500gPlastic bottle
4478200252.5kgPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447825000
Price (EUR)
-
Quantity:
500g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.002 %
Appearance (Color)White
Appearance (Form)Crystals
Nitrite (NO2)=<0.001 %
Phosphate (PO4)=<0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only