N,N-Diisopropylethylenediamine, 97%
N,N-Diisopropylethylenediamine, 97%
N,N-Diisopropylethylenediamine, 97%
Thermo Scientific Chemicals

N,N-Diisopropylethylenediamine, 97%

CAS: 121-05-1 | C8H20N2 | 144.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 448250250
Price (EUR)
62,40
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
62,40
Each
Chemical Identifiers
CAS121-05-1
IUPAC Name(2-aminoethyl)bis(propan-2-yl)amine
Molecular FormulaC8H20N2
InChI KeyCURJNMSGPBXOGK-UHFFFAOYSA-N
SMILESCC(C)N(CCN)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only