4-Bromo-2-furaldehyde, 97%
4-Bromo-2-furaldehyde, 97%
4-Bromo-2-furaldehyde, 97%
Thermo Scientific Chemicals

4-Bromo-2-furaldehyde, 97%

CAS: 21921-76-6 | C5H3BrO2 | 174.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 449440050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS21921-76-6
IUPAC Name4-bromofuran-2-carbaldehyde
Molecular FormulaC5H3BrO2
InChI KeyMRGBBKQOSUHKPF-UHFFFAOYSA-N
SMILESBrC1=COC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)White to brown
Appearance (Form)Crystalline powder and/or chunks
Melting point53°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only