Tris (2-cyanoethyl)phosphine, 95%
Tris (2-cyanoethyl)phosphine, 95%
Tris (2-cyanoethyl)phosphine, 95%
Thermo Scientific Chemicals

Tris (2-cyanoethyl)phosphine, 95%

CAS: 4023-53-4 | C9H12N3P | 193.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
50 g
Catalog number 449710010
Price (EUR)
25,20
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
25,20
Each
Chemical Identifiers
CAS4023-53-4
IUPAC Name3-[bis(2-cyanoethyl)phosphanyl]propanenitrile
Molecular FormulaC9H12N3P
InChI KeyCHZAMJVESILJGH-UHFFFAOYSA-N
SMILESN#CCCP(CCC#N)CCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point95°C to 100°C
Titration Iodimetric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only