2,6-Dichloroindophenol, sodium salt hydrate, 90+%
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
2,6-Dichloroindophenol, sodium salt hydrate, 90+%
Thermo Scientific Chemicals

2,6-Dichloroindophenol, sodium salt hydrate, 90+%

CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 450700050
Price (EUR)
53,70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
53,70
Each
Specifications
Assay Percent Range90+%
CAS1266615-56-8
Chemical Name or Material2, 6-Dichloroindophenol, sodium salt hydrate
Formula Weight290.07
IUPAC Namesodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only