L-Alanine, 99%, specified according to the requirements of Ph.Eur.
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
L-Alanine, 99%, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

L-Alanine, 99%, specified according to the requirements of Ph.Eur.

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 451850010
Price (EUR)
162,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
162,00
Each
Chemical Identifiers
CAS56-41-7
IUPAC Name(2S)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILESC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 % (1 g, 600°C)
Appearance (Color)Colorless to white to almost white
Infrared spectrumConforms
Appearance of solutionreference solution BY6
Specific optical rotation+13.5° to +15.5° (20°C, 589 nm) (c=10, 6N HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only