Benzamide oxime, 97%
Benzamide oxime, 97%
Benzamide oxime, 97%
Thermo Scientific Chemicals

Benzamide oxime, 97%

CAS: 613-92-3 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 453280050
Price (EUR)
97,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
97,00
Each
Chemical Identifiers
CAS613-92-3
IUPAC Name(Z)-N'-hydroxybenzenecarboximidamide
Molecular FormulaC7H8N2O
InChI KeyMXOQNVMDKHLYCZ-UHFFFAOYSA-N
SMILESN\C(=N/O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Infrared spectrumConforms
Melting point73°C to 79°C
HPLC>=94.0 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only