2-(Chloromethyl)-1,3-benzoxazole, 95%
2-(Chloromethyl)-1,3-benzoxazole, 95%
2-(Chloromethyl)-1,3-benzoxazole, 95%
Thermo Scientific Chemicals

2-(Chloromethyl)-1,3-benzoxazole, 95%

CAS: 41014-43-1 | C8H6ClNO | 167.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 453380010
Price (EUR)
123,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
123,00
Each
Chemical Identifiers
CAS41014-43-1
IUPAC Name2-(chloromethyl)-1,3-benzoxazole
Molecular FormulaC8H6ClNO
InChI KeyANRDUCQCZKLSGF-UHFFFAOYSA-N
SMILESClCC1=NC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to orange
GC>=94.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only