Penicillin G sodium salt, 98%
Penicillin G sodium salt, 98%
Penicillin G sodium salt, 98%
Thermo Scientific Chemicals

Penicillin G sodium salt, 98%

Penicillin G sodium salt, CAS # 69-57-8, also called benzylpenicillin, is a β-lactam antibiotic active against gram-positive and gram-negative bacteria.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
45569025025 g
4556900101 g
4556900505 g
Catalog number 455690250
Price (EUR)
91,65
Offre exceptionnelle en ligne
102,00
Save 10,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
91,65
Offre exceptionnelle en ligne
102,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS69-57-8
IUPAC Namesodium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular FormulaC16H17N2NaO4S
InChI KeyFCPVYOBCFFNJFS-LQDWTQKMSA-M
SMILES[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
IdentificationA,B,C,D: Passes tests per BP
Absorbance=<0.10 (280 nm)
Absorbance=<0.10 (325 nm)
2-Ethylhexanoic acid=<0.5 %
Loss on drying=<1.0 % (1 g, 105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Penicillin G sodium salt is the sodium salt of benzylpenicillin, a broad-spectrum β-lactam antibiotic with gram-positive and gram-negative bactericidal activity
• Penicillin G binds and inactivates penicillin-binding-proteins associated with the bacterial cell wall, thus inhibiting cell wall synthesis and resulting in cell lysis
Applications
• Penicillin G can be used as a selection agent in the isolation of microorganisms or in cell culture applications
• It can also be used to prevent infection in research animals
RUO – Research Use Only
Literature references
Livermore, D.M. beta-Lactamases- the Threat Renews. Curr. Protein Pept. Sci. 2009, 10, (5), 397-400.
Demain, A.L. Production of beta-lactam antibiotics and its regulation. Proc. Natl. Sci. Counc. Repub. China B. 1991, 15, (4), 251-65.