Salicylaldazine, 99%
Salicylaldazine, 99%
Salicylaldazine, 99%
Thermo Scientific Chemicals

Salicylaldazine, 99%

CAS: 959-36-4 | C14H12N2O2 | 240.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 456940250
Price (EUR)
173,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
173,00
Each
Chemical Identifiers
CAS959-36-4
IUPAC Name(6E)-6-[(2-{[(1E)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}hydrazin-1-yl)methylidene]cyclohexa-2,4-dien-1-one
Molecular FormulaC14H12N2O2
InChI KeySPEXYYIULCBQJR-WGDLNXRISA-N
SMILESO=C1C=CC=C\C1=C/NN\C=C1/C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only