Indoxyl-beta-D-glucoside, 98%
Indoxyl-beta-D-glucoside, 98%
Indoxyl-beta-D-glucoside, 98%
Thermo Scientific Chemicals

Indoxyl-beta-D-glucoside, 98%

CAS: 487-60-5 | C14H17NO6 | 295.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 458880010
Price (EUR)
233,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
233,00
Each
Chemical Identifiers
CAS487-60-5
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
Molecular FormulaC14H17NO6
InChI KeyXVARCVCWNFACQC-RKQHYHRCSA-N
SMILESOC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Water=<0.5 %
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only