Peptone, from meat (pork gelatine)
Thermo Scientific Chemicals

Peptone, from meat (pork gelatine)

Peptone, from meat (pork gelatine), CAS # 91079-38-8, is a nutrient isolated from meat used to promote the in vitro growth of several species including bacteria and fungi. | CAS: 91079-38-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 458930010
Price (EUR)
86,65
Online Exclusive
96,50
Save 9,85 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
86,65
Online Exclusive
96,50
Save 9,85 (10%)
Each
Chemical Identifiers
CAS91079-38-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
pH6.5 to 7.5 (2 % in water)
Total nitrogen14.0 to 16.0 %
Residue after ign. (sulfated)=<17 %
Microbial contamination=<5000 CFU/g (TAMC)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

● Peptone, from meat, is a growth-promoting nutrient used in bacteria and fungi cultures

Applications

● It is commonly used in animal cell culture.

● Peptone is used in mass scale organism cultivation, and fermentation processes

● Peptone, from meat, is used as a supplement to in vitro culture media to help to increase cell number, specific monoclonal antibody production, and maintain long-term cultures

● This compound has been used as a component in Ax-2 axenic medium to induce reproduction of Dictyostelium discoideum

RUO – Research Use Only

General References:

  1. Jan, D.; Jones, S.; Emery, A.; Al-Rubeai, M. Peptone, a low-cost growth-promoting nutrient for intensive animal cell culture. Cytotechnology. 1994, 16, (1), 17-26.
  2. Amaroli, A.; Gallus, L.; Ferrando, S. Permethrin drastically affects the developmental cycle of the non-target slime mould Dictyostelium discoideum. Chemosphere. 2018, 193, 1-7.