2-Phenylacrylic acid, 94%
2-Phenylacrylic acid, 94%
2-Phenylacrylic acid, 94%
Thermo Scientific Chemicals

2-Phenylacrylic acid, 94%

CAS: 492-38-6 | C9H8O2 | 148.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 459050250
Price (EUR)
156,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
156,00
Each
Chemical Identifiers
CAS492-38-6
IUPAC Name2-phenylprop-2-enoic acid
Molecular FormulaC9H8O2
InChI KeyONPJWQSDZCGSQM-UHFFFAOYSA-N
SMILESOC(=O)C(=C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only