Cefadroxil, 95-105%
Cefadroxil, 95-105%
Cefadroxil, 95-105%
Thermo Scientific Chemicals

Cefadroxil, 95-105%

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4594700505g
4594700101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459470050
Price (EUR)
276,00
Each
Add to cart
Quantity:
5g
Request bulk or custom format
Price (EUR)
276,00
Each
Add to cart
Chemical Identifiers
CAS66592-87-8
IUPAC Name(6R,7R)-7-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate
Molecular FormulaC16H19N3O6S
InChI KeyNBFNMSULHIODTC-CYJZLJNKSA-N
SMILESO.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=C(O)C=C1)C2=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.0 to 6.0 (5% in water)
Specific optical rotation+165° to +178° (20°C, 589 nm) (1% in water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only