Succinic acid disodium salt, for biochemistry, anhydrous
Succinic acid disodium salt, for biochemistry, anhydrous
Succinic acid disodium salt, for biochemistry, anhydrous
Thermo Scientific Chemicals

Succinic acid disodium salt, for biochemistry, anhydrous

CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 459631000
Price (EUR)
26,65
Online Exclusive
28,90
Save 2,25 (8%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
26,65
Online Exclusive
28,90
Save 2,25 (8%)
Each
Chemical Identifiers
CAS150-90-3
IUPAC Namedisodium butanedioate
Molecular FormulaC4H4Na2O4
InChI KeyZDQYSKICYIVCPN-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 280 nm A: =<0.1
UVat 260 nm A: =<0.2
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<10 ppm
Titration with HClO498.5 to 102 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only