3-(1-Pyridinio)-1-propanesulfonate, 99%
3-(1-Pyridinio)-1-propanesulfonate, 99%
3-(1-Pyridinio)-1-propanesulfonate, 99%
Thermo Scientific Chemicals

3-(1-Pyridinio)-1-propanesulfonate, 99%

CAS: 15471-17-7 | C8H11NO3S | 201.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 459670500
Price (EUR)
50,80
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
50,80
Each
Chemical Identifiers
CAS15471-17-7
IUPAC Name1-(3-sulfonatopropyl)pyridin-1-ium
Molecular FormulaC8H11NO3S
InChI KeyREEBJQTUIJTGAL-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CCC[N+]1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Loss on drying=<1 %
HPLC>=98.5 %
Solubility(10% in water) Clear solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only