4-Chloropyrazole, 98%
4-Chloropyrazole, 98%
4-Chloropyrazole, 98%
Thermo Scientific Chemicals

4-Chloropyrazole, 98%

CAS: 15878-00-9 | C3H3ClN2 | 102.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 459920250
Price (EUR)
172,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
172,00
Each
Chemical Identifiers
CAS15878-00-9
IUPAC Name4-chloro-1H-pyrazole
Molecular FormulaC3H3ClN2
InChI KeyBADSZRMNXWLUKO-UHFFFAOYSA-N
SMILESClC1=CNN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Powder or crystalline powder or solid
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only