2-Methyl-4-isothiazolin-3-one, 50% aqueous solution
2-Methyl-4-isothiazolin-3-one, 50% aqueous solution
2-Methyl-4-isothiazolin-3-one, 50% aqueous solution
Thermo Scientific Chemicals

2-Methyl-4-isothiazolin-3-one, 50% aqueous solution

CAS: 2682-20-4 | C4H5NOS | 115.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459970050
Price (EUR)
46,65
Offre exceptionnelle en ligne
52,20
Save 5,55 (11%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
46,65
Offre exceptionnelle en ligne
52,20
Save 5,55 (11%)
Each
Chemical Identifiers
CAS2682-20-4
IUPAC Namehydrogen 2-methyl-2,3-dihydro-1,2-thiazol-3-one chloride
Molecular FormulaC4H6ClNOS
InChI KeySJXPQSRCFCPWQQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN1SC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH5.0 to 7.0 (1 % in water)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Solution
Concentration>=50 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only