Product Image
Thermo Scientific Chemicals

Bile bovine, powder

CAS: 8008-63-7 | C24H40O5 | 408.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460331000
Price (EUR)
55,65
Online exclusive
61,80
Save 6,15 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
55,65
Online exclusive
61,80
Save 6,15 (10%)
Each
Chemical Identifiers
CAS8008-63-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Loss on drying=<5 % (105°C, 2 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only