Artesunate, Thermo Scientific Chemicals
Artesunate, Thermo Scientific Chemicals
Artesunate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Artesunate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4603400505g
46034025025g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460340050
Price (EUR)
36,65
Online exclusive
40,50
Save 3,85 (10%)
Each
Quantity:
5g
Request bulk or custom format
Price (EUR)
36,65
Online exclusive
40,50
Save 3,85 (10%)
Each
Chemical Identifiers
CAS88495-63-0
IUPAC Name4-oxo-4-{[(4S,5R,8S,9R,10R,12R,13R)-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecan-10-yl]oxy}butanoic acid
Molecular FormulaC19H28O8
InChI KeyFIHJKUPKCHIPAT-JQXDXKTESA-N
SMILESC[C@@H]1CC[C@H]2[C@@H](C)[C@@H](OC(=O)CCC(O)=O)O[C@@H]3OC4(C)CC[C@@H]1[C@@]23OO4
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only