Methyl nervonate, 99%, analytical standard for GC
Methyl nervonate, 99%, analytical standard for GC
Methyl nervonate, 99%, analytical standard for GC
Thermo Scientific Chemicals

Methyl nervonate, 99%, analytical standard for GC

CAS: 2733-88-2 | C25H48O2 | 380.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 460375000
Price (EUR)
212,00
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
212,00
Each
Chemical Identifiers
CAS2733-88-2
IUPAC Namemethyl (15Z)-tetracos-15-enoate
Molecular FormulaC25H48O2
InChI KeyAINIZSBLAFHZCP-KHPPLWFESA-N
SMILESCCCCCCCCC=C/CCCCCCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)low melting solid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only