Product Image
Thermo Scientific Chemicals

Artemether, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4607000505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460700050
Price (EUR)
40,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
40,20
Each
Chemical Identifiers
CAS71963-77-4
IUPAC Name(1R,4S,5R,8S,9R,10S,12R,13R)-10-methoxy-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecane
Molecular FormulaC16H26O5
InChI KeySXYIRMFQILZOAM-HVNFFKDJSA-N
SMILESCO[C@H]1O[C@@H]2O[C@@]3(C)CC[C@H]4[C@H](C)CC[C@@H]([C@H]1C)[C@@]24OO3
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point86°C to 90°C
HPLC>=97.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only