Lamotrigine, 98%
Lamotrigine, 98%
Lamotrigine, 98%
Thermo Scientific Chemicals

Lamotrigine, 98%

CAS: 84057-84-1 | C9H7Cl2N5 | 256.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 460850050
Price (EUR)
149,65
Online exclusive
166,00
Save 16,35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
149,65
Online exclusive
166,00
Save 16,35 (10%)
Each
Chemical Identifiers
CAS84057-84-1
IUPAC Name6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
Molecular FormulaC9H7Cl2N5
InChI KeyPYZRQGJRPPTADH-UHFFFAOYSA-N
SMILESNC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C, 3 h)
HPLC>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point216°C to 220°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only