Diflunisal, Thermo Scientific Chemicals
Diflunisal, Thermo Scientific Chemicals
Diflunisal, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diflunisal, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4608900101g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460890010
Price (EUR)
-
Quantity:
1g
Request bulk or custom format
Chemical Identifiers
CAS22494-42-4
IUPAC Name2',4'-difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Molecular FormulaC13H8F2O3
InChI KeyHUPFGZXOMWLGNK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<0.5 % (60°C, 4 H) (vacuum)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only