Lamivudine, 98%
Lamivudine, 98%
Lamivudine, 98%
Thermo Scientific Chemicals

Lamivudine, 98%

CAS: 134678-17-4 | C8H11N3O3S | 229.26 g/mol
Have Questions?
Catalog NumberQuantity
4610900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461090010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS134678-17-4
IUPAC Name4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2-dihydropyrimidin-2-one
Molecular FormulaC8H11N3O3S
InChI KeyJTEGQNOMFQHVDC-NKWVEPMBSA-N
SMILESNC1=NC(=O)N(C=C1)[C@@H]1CS[C@H](CO)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point175°C to 179°C
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only