Product Image
Thermo Scientific Chemicals

Tetrahydroxydiboron, 95%

CAS: 13675-18-8 | B2H4O4 | 89.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 461280050
Price (EUR)
74,65
Online exclusive
83,30
Save 8,65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
74,65
Online exclusive
83,30
Save 8,65 (10%)
Each
Chemical Identifiers
CAS13675-18-8
IUPAC Name(dihydroxyboranyl)boronic acid
Molecular FormulaB2H4O4
InChI KeySKOWZLGOFVSKLB-UHFFFAOYSA-N
SMILESOB(O)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Appearance (Color)White to light brown
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only