Product Image
Thermo Scientific Chemicals

Khellin, 95%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 461800250
Price (EUR)
278,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
278,00
Each
Chemical Identifiers
CAS82-02-0
IUPAC Name4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
Molecular FormulaC14H12O5
InChI KeyHSMPDPBYAYSOBC-UHFFFAOYSA-N
SMILESCOC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point150°C to 154°C
Water=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only