Product Image
Thermo Scientific Chemicals

Parthenolide

CAS: 20554-84-1 | C15H20O3 | 248.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number 462151000
Price (EUR)
134,65
Online exclusive
149,00
Save 14,35
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
134,65
Online exclusive
149,00
Save 14,35
Each
Chemical Identifiers
CAS20554-84-1
IUPAC Name(1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one
Molecular FormulaC15H20O3
InChI KeyKTEXNACQROZXEV-QLIGOWBFSA-N
SMILESC\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point113°C to 117°C
HPLC>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only