7-Hydroxyindole, 95%
7-Hydroxyindole, 95%
7-Hydroxyindole, 95%
Thermo Scientific Chemicals

7-Hydroxyindole, 95%

CAS: 2380-84-9 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462290010
Price (EUR)
364,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
364,00
Each
Chemical Identifiers
CAS2380-84-9
IUPAC Name1H-indol-7-ol
Molecular FormulaC8H7NO
InChI KeyORVPXPKEZLTMNW-UHFFFAOYSA-N
SMILESOC1=C2NC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Solid or crystalline solid
Infrared spectrumConforms
Melting point83°C to 88°C
GC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only