Pazopanib
Thermo Scientific Chemicals

Pazopanib

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462380010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS444731-52-6
IUPAC Name5-({4-[(2,3-dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl}amino)-2-methylbenzene-1-sulfonamide
Molecular FormulaC21H23N7O2S
InChI KeyCUIHSIWYWATEQL-UHFFFAOYSA-N
SMILESCN(C1=CC2=NN(C)C(C)=C2C=C1)C1=NC(NC2=CC(=C(C)C=C2)S(N)(=O)=O)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow or beige
Appearance (Form)Powder
Infrared spectrumConforms
Solubility(0.5 % in DMSO, warmed) Clear solution
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only